Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTGCCATGAACAACTTCCAGGC[C/T]ATCCTGACTCAGGTGAGAATGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600005 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIITA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CIITA - class II, major histocompatibility complex, transactivator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000246.3 | 100 | Intron | NP_000237.2 | |||
NM_001286402.1 | 100 | Intron | NP_001273331.1 | |||
NM_001286403.1 | 100 | Intron | NP_001273332.1 | |||
XM_006720880.3 | 100 | Silent Mutation | GCC,GCT | A,A 27 | XP_006720943.2 | |
XM_011522484.2 | 100 | Silent Mutation | GCC,GCT | A,A 27 | XP_011520786.1 | |
XM_011522485.2 | 100 | Silent Mutation | GCC,GCT | A,A 27 | XP_011520787.1 | |
XM_011522486.2 | 100 | Silent Mutation | GCC,GCT | A,A 27 | XP_011520788.1 | |
XM_011522487.2 | 100 | Intron | XP_011520789.1 | |||
XM_011522488.2 | 100 | Intron | XP_011520790.1 | |||
XM_011522489.2 | 100 | Intron | XP_011520791.1 | |||
XM_011522490.2 | 100 | Intron | XP_011520792.1 | |||
XM_011522491.2 | 100 | Silent Mutation | GCC,GCT | A,A 27 | XP_011520793.1 | |
XM_011522492.2 | 100 | Intron | XP_011520794.1 | |||
XM_011522493.2 | 100 | Intron | XP_011520795.1 | |||
XM_011522494.2 | 100 | UTR 5 | XP_011520796.1 | |||
XM_011522495.2 | 100 | Intron | XP_011520797.1 | |||
XM_011522496.2 | 100 | Intron | XP_011520798.1 |