Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCCACTGGCACCCCCTGGTAG[C/T]GCAGCCACAGGCCTTCCACCACCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614236 | ||||||||||||||||||||
Literature Links: |
SLC38A7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC38A7 - solute carrier family 38 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308384.1 | 1458 | Intron | NP_001295313.1 | |||
NM_018231.2 | 1458 | Missense Mutation | CAC,CGC | H,R 353 | NP_060701.1 | |
XM_017023393.1 | 1458 | Missense Mutation | CAC,CGC | H,R 353 | XP_016878882.1 | |
XM_017023394.1 | 1458 | Intron | XP_016878883.1 | |||
XM_017023395.1 | 1458 | Intron | XP_016878884.1 | |||
XM_017023396.1 | 1458 | Intron | XP_016878885.1 | |||
XM_017023397.1 | 1458 | Intron | XP_016878886.1 | |||
XM_017023398.1 | 1458 | Intron | XP_016878887.1 | |||
XM_017023399.1 | 1458 | Intron | XP_016878888.1 |