Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCGTCACGATGTGCTGGAGGGTG[C/T]GGGACGGCTGCAGGCCCAGGGCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606978 MIM: 600760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COG7 - component of oligomeric golgi complex 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153603.3 | 2389 | Missense Mutation | CAC,CGC | H,R 731 | NP_705831.1 | |
XM_017023870.1 | 2389 | Missense Mutation | CAC,CGC | H,R 666 | XP_016879359.1 |
SCNN1B - sodium channel epithelial 1 beta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |