Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTTCCCCCTTTGGCAGGTGGT[A/G]CCGGCAGCACCAGGCCCAGAGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602622 MIM: 603022 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNASE1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNASE1L2 - deoxyribonuclease I-like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
E4F1 - E4F transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288776.1 | 511 | Silent Mutation | GTA,GTG | V,V 105 | NP_001275705.1 | |
NM_001288778.1 | 511 | Silent Mutation | GTA,GTG | V,V 105 | NP_001275707.1 | |
NM_004424.4 | 511 | Silent Mutation | GTA,GTG | V,V 105 | NP_004415.3 | |
XM_005255155.1 | 511 | Silent Mutation | GTA,GTG | V,V 158 | XP_005255212.1 | |
XM_005255156.1 | 511 | Silent Mutation | GTA,GTG | V,V 158 | XP_005255213.1 | |
XM_006720858.1 | 511 | Silent Mutation | GTA,GTG | V,V 105 | XP_006720921.1 | |
XM_011522402.1 | 511 | Silent Mutation | GTA,GTG | V,V 158 | XP_011520704.1 | |
XM_011522403.1 | 511 | Silent Mutation | GTA,GTG | V,V 145 | XP_011520705.1 | |
XM_017023010.1 | 511 | Silent Mutation | GTA,GTG | V,V 158 | XP_016878499.1 | |
XM_017023011.1 | 511 | Silent Mutation | GTA,GTG | V,V 145 | XP_016878500.1 |