Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCCAGCTCCAGGAGGGAGAATG[G/T]GAGAGGGGCAAGGAGGGAACGAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616167 MIM: 614709 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCUN1D3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCUN1D3 - defective in cullin neddylation 1 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173475.3 | 166 | Intron | NP_775746.1 |
LYRM1 - LYR motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128301.2 | 166 | Intron | NP_001121773.1 | |||
NM_001128302.2 | 166 | Intron | NP_001121774.1 | |||
NM_001302835.1 | 166 | Intron | NP_001289764.1 | |||
NM_001302836.1 | 166 | UTR 5 | NP_001289765.1 | |||
NM_020424.4 | 166 | UTR 5 | NP_065157.1 | |||
XM_005255444.1 | 166 | Intron | XP_005255501.1 | |||
XM_011545900.2 | 166 | Intron | XP_011544202.1 | |||
XM_011545901.2 | 166 | Intron | XP_011544203.1 | |||
XM_011545902.1 | 166 | UTR 5 | XP_011544204.1 | |||
XM_011545908.2 | 166 | Intron | XP_011544210.1 | |||
XM_017023475.1 | 166 | Missense Mutation | TGG,TGT | W,C 181 | XP_016878964.1 | |
XM_017023476.1 | 166 | Missense Mutation | TGG,TGT | W,C 181 | XP_016878965.1 | |
XM_017023477.1 | 166 | Missense Mutation | TGG,TGT | W,C 181 | XP_016878966.1 | |
XM_017023478.1 | 166 | Intron | XP_016878967.1 | |||
XM_017023479.1 | 166 | Intron | XP_016878968.1 | |||
XM_017023480.1 | 166 | UTR 5 | XP_016878969.1 | |||
XM_017023481.1 | 166 | Intron | XP_016878970.1 | |||
XM_017023482.1 | 166 | Intron | XP_016878971.1 |