Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTAGATTTTCTTAAATAGTTAA[C/T]CTACATTTAAAGCAGAGGACCTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CMC2 - C-X9-C motif containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020188.3 | Intron | NP_064573.1 | ||||
XM_017023461.1 | Intron | XP_016878950.1 | ||||
XM_017023462.1 | Intron | XP_016878951.1 | ||||
XM_017023463.1 | Intron | XP_016878952.1 | ||||
XM_017023464.1 | Intron | XP_016878953.1 | ||||
XM_017023465.1 | Intron | XP_016878954.1 | ||||
XM_017023466.1 | Intron | XP_016878955.1 | ||||
XM_017023467.1 | Intron | XP_016878956.1 | ||||
XM_017023468.1 | Intron | XP_016878957.1 | ||||
XM_017023469.1 | Intron | XP_016878958.1 | ||||
XM_017023470.1 | Intron | XP_016878959.1 |