Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCCTCTCTCTCCACAGAGGGA[A/G]ATAACGCTGTGCTGCAGTGCCTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 107265 MIM: 611869 | ||||||||||||||||||||
Literature Links: |
CD19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD19 - CD19 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178098.1 | 156 | Missense Mutation | AAT,GAT | N,D 32 | NP_001171569.1 | |
NM_001770.5 | 156 | Missense Mutation | AAT,GAT | N,D 32 | NP_001761.3 | |
XM_006721103.3 | 156 | Intron | XP_006721166.1 | |||
XM_011545981.1 | 156 | Missense Mutation | AAT,GAT | N,D 32 | XP_011544283.1 | |
XM_017023893.1 | 156 | Intron | XP_016879382.1 |
RABEP2 - rabaptin, RAB GTPase binding effector protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |