Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATATCTAAGCCCTGTGAGCCTC[A/G]TTTCCTTATCAGAGGCCTGGGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605349 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NARF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NARF - nuclear prelamin A recognition factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038618.2 | Intron | NP_001033707.1 | ||||
NM_001083608.1 | Intron | NP_001077077.1 | ||||
NM_012336.3 | Intron | NP_036468.1 | ||||
NM_031968.2 | Intron | NP_114174.1 | ||||
XM_005256340.3 | Intron | XP_005256397.1 | ||||
XM_006722278.3 | Intron | XP_006722341.1 | ||||
XM_011523541.2 | Intron | XP_011521843.1 | ||||
XM_011523543.2 | Intron | XP_011521845.1 | ||||
XM_011523544.2 | Intron | XP_011521846.1 | ||||
XM_017024449.1 | Intron | XP_016879938.1 | ||||
XM_017024450.1 | Intron | XP_016879939.1 |