Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTCATCCACCAAGCCAGCCTTG[A/G]CCACATTGGCCTTGTAGTAAGCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613422 | ||||||||||||||||||||
Literature Links: |
ATP5H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP5H - ATP synthase, H+ transporting, mitochondrial Fo complex subunit D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003785.1 | Intron | NP_001003785.1 | ||||
NM_006356.2 | Intron | NP_006347.1 |
KCTD2 - potassium channel tetramerization domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015353.2 | Intron | NP_056168.1 | ||||
XM_011524575.2 | Intron | XP_011522877.1 |