Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCCCCTAGAAAGAGGTGGTGC[A/G]GAGTACCAGGCTGTGCTGCCCGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610008 MIM: 188830 MIM: 609224 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARSG - arylsulfatase G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267727.1 | 2323 | Silent Mutation | GCA,GCG | A,A 473 | NP_001254656.1 | |
NM_014960.4 | 2323 | Silent Mutation | GCA,GCG | A,A 473 | NP_055775.2 | |
XM_005257170.3 | 2323 | Silent Mutation | GCA,GCG | A,A 473 | XP_005257227.1 | |
XM_005257172.3 | 2323 | Intron | XP_005257229.1 | |||
XM_006721777.3 | 2323 | Silent Mutation | GCA,GCG | A,A 500 | XP_006721840.2 | |
XM_006721779.3 | 2323 | Intron | XP_006721842.1 | |||
XM_011524535.2 | 2323 | Silent Mutation | GCA,GCG | A,A 500 | XP_011522837.1 | |
XM_011524536.2 | 2323 | Silent Mutation | GCA,GCG | A,A 500 | XP_011522838.1 | |
XM_011524537.1 | 2323 | Silent Mutation | GCA,GCG | A,A 500 | XP_011522839.1 | |
XM_011524538.2 | 2323 | Silent Mutation | GCA,GCG | A,A 500 | XP_011522840.1 | |
XM_011524540.2 | 2323 | Silent Mutation | GCA,GCG | A,A 499 | XP_011522842.1 | |
XM_011524541.2 | 2323 | Intron | XP_011522843.1 | |||
XM_011524542.2 | 2323 | Intron | XP_011522844.1 | |||
XM_011524543.2 | 2323 | Intron | XP_011522845.1 | |||
XM_011524544.2 | 2323 | Intron | XP_011522846.1 | |||
XM_011524545.2 | 2323 | UTR 3 | XP_011522847.1 | |||
XM_011524546.2 | 2323 | Silent Mutation | GCA,GCG | A,A 251 | XP_011522848.1 | |
XM_017024360.1 | 2323 | Silent Mutation | GCA,GCG | A,A 500 | XP_016879849.1 | |
XM_017024361.1 | 2323 | Silent Mutation | GCA,GCG | A,A 473 | XP_016879850.1 | |
XM_017024362.1 | 2323 | Silent Mutation | GCA,GCG | A,A 473 | XP_016879851.1 | |
XM_017024363.1 | 2323 | Silent Mutation | GCA,GCG | A,A 473 | XP_016879852.1 | |
XM_017024364.1 | 2323 | Silent Mutation | GCA,GCG | A,A 473 | XP_016879853.1 | |
XM_017024365.1 | 2323 | Silent Mutation | GCA,GCG | A,A 473 | XP_016879854.1 | |
XM_017024366.1 | 2323 | Intron | XP_016879855.1 | |||
XM_017024367.1 | 2323 | Intron | XP_016879856.1 | |||
XM_017024368.1 | 2323 | UTR 3 | XP_016879857.1 |
MIR635 - microRNA 635 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKAR1A - protein kinase cAMP-dependent type I regulatory subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276289.1 | 2323 | Intron | NP_001263218.1 | |||
NM_001276290.1 | 2323 | Intron | NP_001263219.1 | |||
NM_001278433.1 | 2323 | Intron | NP_001265362.1 | |||
NM_002734.4 | 2323 | Intron | NP_002725.1 | |||
NM_212471.2 | 2323 | Intron | NP_997636.1 | |||
NM_212472.2 | 2323 | Intron | NP_997637.1 | |||
XM_011524983.2 | 2323 | Intron | XP_011523285.1 | |||
XM_011524984.2 | 2323 | Intron | XP_011523286.1 | |||
XM_011524985.2 | 2323 | Intron | XP_011523287.1 |
WIPI1 - WD repeat domain, phosphoinositide interacting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320772.1 | 2323 | Intron | NP_001307701.1 | |||
NM_017983.6 | 2323 | Intron | NP_060453.3 | |||
XM_017024808.1 | 2323 | Intron | XP_016880297.1 |