Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTACTATCAGGAACTTAGAAGGC[C/T]TCCAGAATCTTCACAGTCTCTATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611825 MIM: 614966 | ||||||||||||||||||||
Literature Links: |
LRRC46 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC46 - leucine rich repeat containing 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033413.3 | 556 | Missense Mutation | CTC,TTC | L,F 65 | NP_219481.1 | |
XM_005257775.4 | 556 | Missense Mutation | CTC,TTC | L,F 74 | XP_005257832.1 |
MRPL10 - mitochondrial ribosomal protein L10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCRN2 - secernin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |