Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGCCCTTTACCACTCGACGGTC[A/G]AGAACTTTTTCCACCACATATTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604511 | ||||||||||||||||||||
Literature Links: |
CBX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CBX1 - chromobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127228.1 | 562 | Intron | NP_001120700.1 | |||
NM_006807.4 | 562 | Silent Mutation | CTC,CTT | L,L 27 | NP_006798.1 |