Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGAGGAGTAACCTGACAAAGGT[A/G]GAAATACAAATGCTGTGATGTAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609835 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYCBPAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYCBPAP - MYCBP associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032133.4 | Intron | NP_115509.4 | ||||
XM_005257725.3 | Intron | XP_005257782.1 | ||||
XM_005257726.2 | Intron | XP_005257783.1 | ||||
XM_011525342.1 | Intron | XP_011523644.1 | ||||
XM_011525343.1 | Intron | XP_011523645.1 | ||||
XM_017025205.1 | Intron | XP_016880694.1 |