Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGCTGTGTACGCTGGGCCGGCGC[A/G]CGAAGGGTGCCACGATGCCAAAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609878 MIM: 605550 | ||||||||||||||||||||
Literature Links: |
MED9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MED9 - mediator complex subunit 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASD1 - ras related dexamethasone induced 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199989.1 | 889 | UTR 3 | NP_001186918.1 | |||
NM_016084.4 | 889 | Missense Mutation | GCG,GTG | A,V 250 | NP_057168.1 |