Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGACGGGGCGAGGAGGCTCACC[A/G]GAATTCTTGTGTTTTTCACTTCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606363 MIM: 139391 | ||||||||||||||||||||
Literature Links: |
ABI3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABI3 - ABI family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNGT2 - G protein subunit gamma transducin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198754.1 | 373 | Missense Mutation | CCG,CTG | P,L 28 | NP_001185683.1 | |
NM_001198755.1 | 373 | Missense Mutation | CCG,CTG | P,L 28 | NP_001185684.1 | |
NM_001198756.1 | 373 | Missense Mutation | CCG,CTG | P,L 28 | NP_001185685.1 | |
NM_031498.2 | 373 | Missense Mutation | CCG,CTG | P,L 28 | NP_113686.1 |