Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGCTTCTGGGGATCCTGTTCTG[C/G]GGAGCCACAGCCAGAGACTGGCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607873 MIM: 610791 | ||||||||||||||||||||
Literature Links: |
SCARF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCARF1 - scavenger receptor class F member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003693.3 | 1391 | Missense Mutation | CCA,GCA | P,A 800 | NP_003684.2 | |
NM_145350.2 | 1391 | UTR 3 | NP_663325.1 | |||
XM_017025256.1 | 1391 | Missense Mutation | CCA,GCA | P,A 389 | XP_016880745.1 |
SLC43A2 - solute carrier family 43 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |