Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTGTATAGGACTGGAGGACTGT[C/T]CTGGGAAATCAGAAGAGCATCACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614955 | ||||||||||||||||||||
Literature Links: |
SLFN12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLFN12 - schlafen family member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289009.1 | 1857 | Missense Mutation | AAC,GAC | N,D 450 | NP_001275938.1 | |
NM_018042.4 | 1857 | Missense Mutation | AAC,GAC | N,D 450 | NP_060512.3 | |
XM_005257995.4 | 1857 | Missense Mutation | AAC,GAC | N,D 450 | XP_005258052.1 | |
XM_011524967.2 | 1857 | Missense Mutation | AAC,GAC | N,D 450 | XP_011523269.1 | |
XM_017024809.1 | 1857 | Missense Mutation | AAC,GAC | N,D 450 | XP_016880298.1 | |
XM_017024810.1 | 1857 | Missense Mutation | AAC,GAC | N,D 450 | XP_016880299.1 | |
XM_017024811.1 | 1857 | UTR 3 | XP_016880300.1 |