Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTTACATTACCATCAATGTCA[A/T]CAATTTTACTTTCTTCAATATGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605882 | ||||||||||||||||||||
Literature Links: |
BRIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRIP1 - BRCA1 interacting protein C-terminal helicase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032043.2 | 4417 | Missense Mutation | GAT,GTT | D,V 1207 | NP_114432.2 | |
XM_011525332.2 | 4417 | Missense Mutation | GAT,GTT | D,V 1227 | XP_011523634.1 | |
XM_011525333.2 | 4417 | Missense Mutation | GAT,GTT | D,V 1227 | XP_011523635.1 | |
XM_011525334.2 | 4417 | Missense Mutation | GAT,GTT | D,V 1227 | XP_011523636.1 | |
XM_011525335.2 | 4417 | Missense Mutation | GAT,GTT | D,V 1207 | XP_011523637.1 | |
XM_011525336.2 | 4417 | Missense Mutation | GAT,GTT | D,V 1187 | XP_011523638.1 | |
XM_011525337.2 | 4417 | Missense Mutation | GAT,GTT | D,V 1160 | XP_011523639.1 | |
XM_011525338.2 | 4417 | Missense Mutation | GAT,GTT | D,V 1066 | XP_011523640.1 | |
XM_011525339.2 | 4417 | Intron | XP_011523641.1 | |||
XM_011525340.2 | 4417 | Intron | XP_011523642.1 | |||
XM_011525341.2 | 4417 | Intron | XP_011523643.1 | |||
XM_017025200.1 | 4417 | Missense Mutation | GAT,GTT | D,V 1046 | XP_016880689.1 | |
XM_017025201.1 | 4417 | Missense Mutation | GAT,GTT | D,V 1046 | XP_016880690.1 | |
XM_017025202.1 | 4417 | Missense Mutation | GAT,GTT | D,V 589 | XP_016880691.1 | |
XM_017025203.1 | 4417 | Missense Mutation | GAT,GTT | D,V 589 | XP_016880692.1 |