Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCCAAAGTGGGCCTGGGCACAG[A/C]CGAGGGCACCTCCACCCCCAGGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612341 MIM: 604885 | ||||||||||||||||||||
Literature Links: |
GGT6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GGT6 - gamma-glutamyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122890.2 | 844 | Missense Mutation | GCT,TCT | A,S 289 | NP_001116362.1 | |
NM_001288702.1 | 844 | Missense Mutation | GCT,TCT | A,S 295 | NP_001275631.1 | |
NM_001288703.1 | 844 | Intron | NP_001275632.1 | |||
NM_001288704.1 | 844 | Intron | NP_001275633.1 | |||
NM_153338.3 | 844 | Missense Mutation | GCT,TCT | A,S 257 | NP_699169.2 |
MYBBP1A - MYB binding protein 1a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |