Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGACAAAGCAAATTTCATGTCC[C/G]TCCTACACCAATAAAGAGAAAAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GLOD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLOD4 - glyoxalase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016080.3 | 845 | Silent Mutation | GAC,GAG | D,E 249 | NP_057164.3 | |
XM_006721540.2 | 845 | Silent Mutation | GAC,GAG | D,E 240 | XP_006721603.1 | |
XM_017024711.1 | 845 | Silent Mutation | GAC,GAG | D,E 240 | XP_016880200.1 | |
XM_017024712.1 | 845 | Missense Mutation | GAC,GAG | D,E 193 | XP_016880201.1 |