Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTGGTGCCAATGCTCTTCTCTG[A/C]GGCTCTCAGTGCAGCTGCCAGGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612185 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASKIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASKIN2 - CASK interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142643.2 | 4071 | Missense Mutation | NP_001136115.1 | |||
NM_020753.4 | 4071 | Missense Mutation | NP_065804.2 |
MIR6785 - microRNA 6785 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM94 - transmembrane protein 94 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |