Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCACAAAGCTCCAGGAGCGTC[C/T]GAGGGCAGGTTTGAGCCTCCCGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602954 | ||||||||||||||||||||
Literature Links: |
RAD51D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAD51D - RAD51 paralog D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142571.1 | 1109 | Missense Mutation | AGA,GGA | R,G 285 | NP_001136043.1 | |
NM_002878.3 | 1109 | Missense Mutation | AGA,GGA | R,G 265 | NP_002869.3 | |
NM_133629.2 | 1109 | Missense Mutation | AGA,GGA | R,G 153 | NP_598332.1 |
RAD51L3-RFFL - RAD51L3-RFFL readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |