Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCCTTTTAAATGGATCAAGCCA[A/G]GACAGAATGTTTGAGACAATGGCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604026 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOSR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GOSR1 - golgi SNAP receptor complex member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007024.1 | 456 | UTR 5 | NP_001007025.1 | |||
NM_001007025.1 | 456 | Silent Mutation | CAA,CAG | Q,Q 61 | NP_001007026.1 | |
NM_004871.2 | 456 | Silent Mutation | CAA,CAG | Q,Q 63 | NP_004862.1 | |
XM_005258070.4 | 456 | Silent Mutation | CAA,CAG | Q,Q 113 | XP_005258127.1 | |
XM_005258071.4 | 456 | Silent Mutation | CAA,CAG | Q,Q 111 | XP_005258128.1 | |
XM_005258072.2 | 456 | UTR 5 | XP_005258129.1 | |||
XM_017025374.1 | 456 | Silent Mutation | CAA,CAG | Q,Q 113 | XP_016880863.1 | |
XM_017025375.1 | 456 | Silent Mutation | CAA,CAG | Q,Q 63 | XP_016880864.1 | |
XM_017025376.1 | 456 | Silent Mutation | CAA,CAG | Q,Q 61 | XP_016880865.1 | |
XM_017025377.1 | 456 | Silent Mutation | CAA,CAG | Q,Q 113 | XP_016880866.1 |