Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAGCCTCCCAGGCAGCCGGGTC[A/G]TGCTTGAAGAGGGAGTTGGTGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173325 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JUP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JUP - junction plakoglobin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002230.2 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | NP_002221.1 | |
NM_021991.2 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | NP_068831.1 | |
XM_006721873.2 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | XP_006721936.1 | |
XM_006721874.2 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | XP_006721937.1 | |
XM_006721875.1 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | XP_006721938.1 | |
XM_006721878.1 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | XP_006721941.1 | |
XM_011524753.2 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | XP_011523055.1 | |
XM_011524755.1 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | XP_011523057.1 | |
XM_011524756.1 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | XP_011523058.1 | |
XM_011524757.2 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | XP_011523059.1 | |
XM_011524758.1 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | XP_011523060.1 | |
XM_017024588.1 | 2408 | Silent Mutation | CAC,CAT | H,H 692 | XP_016880077.1 | |
XM_017024589.1 | 2408 | Silent Mutation | CAC,CAT | H,H 692 | XP_016880078.1 | |
XM_017024590.1 | 2408 | Silent Mutation | CAC,CAT | H,H 675 | XP_016880079.1 |