Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATCAGAATCTGGACCAGACAAC[A/G]AACTGAGCATCCTGGGCAAGGTAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611358 | ||||||||||||||||||||
Literature Links: |
DPRXP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPRXP4 - divergent-paired related homeobox pseudogene 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF135 - ring finger protein 135 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184992.1 | 818 | Missense Mutation | AAA,GAA | K,E 166 | NP_001171921.1 | |
NM_032322.3 | 818 | Missense Mutation | AAA,GAA | K,E 166 | NP_115698.3 | |
NM_197939.1 | 818 | Missense Mutation | AAA,GAA | K,E 166 | NP_922921.1 | |
XM_005258043.4 | 818 | UTR 5 | XP_005258100.1 | |||
XM_017025223.1 | 818 | UTR 5 | XP_016880712.1 |