Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATAGAAGATACTACTTACCTTC[G/T]TCCAGCTTAAAAAAAGATTGTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612508 MIM: 612503 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA10 - ATP binding cassette subfamily A member 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ABCA5 - ATP binding cassette subfamily A member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018672.4 | 5979 | Missense Mutation | GAA,GAC | E,D 1587 | NP_061142.2 | |
NM_172232.3 | 5979 | Missense Mutation | GAA,GAC | E,D 1587 | NP_758424.1 |