Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTTGCTCAGGGTCCATACCACAG[A/G]AGCGGAGGGTGGTGAGTGGGTAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608385 | ||||||||||||||||||||
Literature Links: |
TNS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNS4 - tensin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032865.5 | 2192 | Missense Mutation | TCC,TTC | S,F 651 | NP_116254.4 | |
XM_005257744.1 | 2192 | Missense Mutation | TCC,TTC | S,F 650 | XP_005257801.1 | |
XM_017025236.1 | 2192 | Missense Mutation | TCC,TTC | S,F 625 | XP_016880725.1 | |
XM_017025237.1 | 2192 | Intron | XP_016880726.1 |