Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGGGCGGGCCACTCGGGCTGCT[A/G]TGGCCACACAGGAGCAGGGGAATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603333 MIM: 602297 | ||||||||||||||||||||
Literature Links: |
DNAH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAH2 - dynein axonemal heavy chain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303270.1 | 174 | Missense Mutation | ATG,GTG | M,V 30 | NP_001290199.1 | |
NM_020877.3 | 174 | Missense Mutation | ATG,GTG | M,V 30 | NP_065928.2 | |
XM_011523663.1 | 174 | Missense Mutation | ATG,GTG | M,V 30 | XP_011521965.1 | |
XM_011523664.2 | 174 | Missense Mutation | ATG,GTG | M,V 30 | XP_011521966.1 | |
XM_011523666.1 | 174 | Missense Mutation | ATG,GTG | M,V 30 | XP_011521968.1 | |
XM_011523667.2 | 174 | Missense Mutation | ATG,GTG | M,V 30 | XP_011521969.1 | |
XM_011523668.1 | 174 | Intron | XP_011521970.1 | |||
XM_011523669.2 | 174 | Intron | XP_011521971.1 | |||
XM_011523670.2 | 174 | Missense Mutation | ATG,GTG | M,V 30 | XP_011521972.1 | |
XM_017024218.1 | 174 | Intron | XP_016879707.1 | |||
XM_017024219.1 | 174 | Missense Mutation | ATG,GTG | M,V 30 | XP_016879708.1 |
EFNB3 - ephrin B3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |