Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGGTCACTTGTGGTAGAATCG[C/T]GGGTTCTGGCTGTGTTGGATGAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600187 MIM: 601935 MIM: 615865 | ||||||||||||||||||||
Literature Links: |
EIF5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF5A - eukaryotic translation initiation factor 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143760.1 | 1042 | Intron | NP_001137232.1 | |||
NM_001143761.1 | 1042 | Intron | NP_001137233.1 | |||
NM_001143762.1 | 1042 | Intron | NP_001137234.1 | |||
NM_001970.4 | 1042 | Intron | NP_001961.1 | |||
XM_005256509.2 | 1042 | Intron | XP_005256566.1 | |||
XM_011523710.2 | 1042 | Intron | XP_011522012.1 | |||
XM_011523711.2 | 1042 | Intron | XP_011522013.1 | |||
XM_011523712.2 | 1042 | Intron | XP_011522014.1 | |||
XM_011523713.2 | 1042 | Intron | XP_011522015.1 | |||
XM_017024300.1 | 1042 | Intron | XP_016879789.1 | |||
XM_017024301.1 | 1042 | Intron | XP_016879790.1 |
GPS2 - G protein pathway suppressor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004489.4 | 1042 | Silent Mutation | CCA,CCG | P,P 322 | NP_004480.1 |
NEURL4 - neuralized E3 ubiquitin protein ligase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |