Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCAGAACCTGCTCATCAATGAG[G/T]TGGGTGCCATCAAGCTGGCTGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123828 MIM: 601590 MIM: 613130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK3 - cyclin dependent kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258.2 | 488 | Silent Mutation | GTG,TTG | V,L 138 | NP_001249.1 |
EVPL - envoplakin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEN1 - TEN1 CST complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEN1-CDK3 - TEN1-CDK3 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |