Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTGGTGAATTTTTGAAACTGGA[C/T]AGAGAAAGAGCCAAGAACAAAATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603352 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFMID PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AFMID - arylformamidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BIRC5 - baculoviral IAP repeat containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012270.1 | 436 | Intron | NP_001012270.1 | |||
NM_001012271.1 | 436 | Silent Mutation | GAC,GAT | D,D 128 | NP_001012271.1 | |
NM_001168.2 | 436 | Silent Mutation | GAC,GAT | D,D 105 | NP_001159.2 |