Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGACCCTGCCTGCCTGTGTCTCAG[A/G]TCTCACCAACATCCTGGCTTCCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 106180 | ||||||||||||||||||||
Literature Links: |
ACE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACE - angiotensin I converting enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000789.3 | 546 | Missense Mutation | GAT,GGT | D,G 171 | NP_000780.1 | |
NM_001178057.1 | 546 | Intron | NP_001171528.1 | |||
NM_152830.2 | 546 | Intron | NP_690043.1 | |||
XM_006721737.3 | 546 | Intron | XP_006721800.2 |