Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTGTTAGTTGCCAGATACCCTT[C/T]CTGGCAATGGCAAACCCTGCGTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612816 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBTD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MBTD1 - mbt domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017643.2 | Intron | NP_060113.2 | ||||
XM_005257467.4 | Intron | XP_005257524.1 | ||||
XM_005257468.4 | Intron | XP_005257525.1 | ||||
XM_011524922.2 | Intron | XP_011523224.1 | ||||
XM_011524923.2 | Intron | XP_011523225.1 | ||||
XM_011524924.2 | Intron | XP_011523226.1 | ||||
XM_011524925.1 | Intron | XP_011523227.1 | ||||
XM_011524926.2 | Intron | XP_011523228.1 | ||||
XM_011524928.2 | Intron | XP_011523230.1 | ||||
XM_011524929.2 | Intron | XP_011523231.1 | ||||
XM_011524930.1 | Intron | XP_011523232.1 | ||||
XM_011524931.2 | Intron | XP_011523233.1 | ||||
XM_011524933.2 | Intron | XP_011523235.1 | ||||
XM_011524934.2 | Intron | XP_011523236.1 | ||||
XM_011524935.2 | Intron | XP_011523237.1 | ||||
XM_011524936.1 | Intron | XP_011523238.1 | ||||
XM_017024782.1 | Intron | XP_016880271.1 |
UTP18 - UTP18, small subunit processome component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016001.2 | Intron | NP_057085.2 | ||||
XM_006721930.3 | Intron | XP_006721993.2 | ||||
XM_011524870.2 | Intron | XP_011523172.1 |