Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGATGGAAATTTCATATTCTTCC[G/T]TAAAAGCTAAAATTGTAATTCAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615047 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TANC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TANC2 - tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025185.3 | Intron | NP_079461.2 | ||||
XM_005257203.4 | Intron | XP_005257260.1 | ||||
XM_006721806.3 | Intron | XP_006721869.1 | ||||
XM_006721807.3 | Intron | XP_006721870.1 | ||||
XM_006721810.3 | Intron | XP_006721873.1 | ||||
XM_006721811.3 | Intron | XP_006721874.1 | ||||
XM_011524597.2 | Intron | XP_011522899.1 | ||||
XM_011524598.2 | Intron | XP_011522900.1 | ||||
XM_011524599.1 | Intron | XP_011522901.1 | ||||
XM_011524600.2 | Intron | XP_011522902.1 | ||||
XM_011524601.2 | Intron | XP_011522903.1 | ||||
XM_011524602.2 | Intron | XP_011522904.1 | ||||
XM_017024429.1 | Intron | XP_016879918.1 | ||||
XM_017024430.1 | Intron | XP_016879919.1 | ||||
XM_017024431.1 | Intron | XP_016879920.1 | ||||
XM_017024432.1 | Intron | XP_016879921.1 |