Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGCTCTTCCGCCTGGGAAGGCC[A/G]CTTCCAGTCGGCCCGGGTCAAGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600019 MIM: 615047 | ||||||||||||||||||||
Literature Links: |
CYB561 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYB561 - cytochrome b561 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017916.1 | 848 | Missense Mutation | CGG,TGG | R,W 226 | NP_001017916.1 | |
NM_001017917.1 | 848 | Missense Mutation | CGG,TGG | R,W 226 | NP_001017917.1 | |
NM_001915.3 | 848 | Missense Mutation | CGG,TGG | R,W 226 | NP_001906.3 | |
XM_005257091.1 | 848 | Missense Mutation | CGG,TGG | R,W 226 | XP_005257148.1 | |
XM_017024260.1 | 848 | Missense Mutation | CGG,TGG | R,W 233 | XP_016879749.1 | |
XM_017024261.1 | 848 | UTR 3 | XP_016879750.1 | |||
XM_017024262.1 | 848 | Missense Mutation | CGG,TGG | R,W 293 | XP_016879751.1 |
TANC2 - tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |