Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCATGGAGGAGCGCCGGATGTGC[C/T]GACAGGAAGACCACTGCCGGAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600242 | ||||||||||||||||||||
Literature Links: |
CCR7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCR7 - C-C motif chemokine receptor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301714.1 | 1157 | Missense Mutation | NP_001288643.1 | |||
NM_001301716.1 | 1157 | Missense Mutation | NP_001288645.1 | |||
NM_001301717.1 | 1157 | Missense Mutation | NP_001288646.1 | |||
NM_001301718.1 | 1157 | Missense Mutation | NP_001288647.1 | |||
NM_001838.3 | 1157 | Missense Mutation | NP_001829.1 |