Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATAACTTCTCCTCGCAGCGCCCC[A/G]CCGTCCTCAGAATAACCTGGAAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610008 MIM: 188830 MIM: 609224 | ||||||||||||||||||||
Literature Links: |
ARSG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR635 - microRNA 635 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKAR1A - protein kinase cAMP-dependent type I regulatory subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276289.1 | 1312 | Intron | NP_001263218.1 | |||
NM_001276290.1 | 1312 | Intron | NP_001263219.1 | |||
NM_001278433.1 | 1312 | Intron | NP_001265362.1 | |||
NM_002734.4 | 1312 | Intron | NP_002725.1 | |||
NM_212471.2 | 1312 | Intron | NP_997636.1 | |||
NM_212472.2 | 1312 | Intron | NP_997637.1 | |||
XM_011524983.2 | 1312 | Intron | XP_011523285.1 | |||
XM_011524984.2 | 1312 | Intron | XP_011523286.1 | |||
XM_011524985.2 | 1312 | Intron | XP_011523287.1 |
WIPI1 - WD repeat domain, phosphoinositide interacting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320772.1 | 1312 | Silent Mutation | GGC,GGT | G,G 321 | NP_001307701.1 | |
NM_017983.6 | 1312 | Silent Mutation | GGC,GGT | G,G 403 | NP_060453.3 | |
XM_017024808.1 | 1312 | Silent Mutation | GGC,GGT | G,G 275 | XP_016880297.1 |