Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGTTTTGGAAGACTCAACACCT[C/T]CTCCATCTTTTGCTGTTGATAATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602552 MIM: 603616 | ||||||||||||||||||||
Literature Links: |
NUP88 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUP88 - nucleoporin 88 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320653.1 | 2270 | Missense Mutation | AAG,GAG | K,E 711 | NP_001307582.1 | |
NM_002532.5 | 2270 | Missense Mutation | AAG,GAG | K,E 695 | NP_002523.2 | |
XM_011523893.2 | 2270 | Missense Mutation | AAG,GAG | K,E 580 | XP_011522195.1 | |
XM_017024694.1 | 2270 | Missense Mutation | AAG,GAG | K,E 564 | XP_016880183.1 |
RABEP1 - rabaptin, RAB GTPase binding effector protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083585.2 | 2270 | Intron | NP_001077054.1 | |||
NM_001291581.1 | 2270 | Intron | NP_001278510.1 | |||
NM_001291582.1 | 2270 | Intron | NP_001278511.1 | |||
NM_004703.5 | 2270 | Intron | NP_004694.2 | |||
XM_011524060.1 | 2270 | Intron | XP_011522362.1 | |||
XM_017025294.1 | 2270 | Intron | XP_016880783.1 | |||
XM_017025295.1 | 2270 | Intron | XP_016880784.1 |