Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAACTTGGAGAATGGAAAAGAAC[C/T]ATTGAAGCTTGAGAGAAAAGCCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF286A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF286A - zinc finger protein 286A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130842.1 | Intron | NP_001124314.1 | ||||
NM_001288642.1 | Intron | NP_001275571.1 | ||||
NM_001288643.1 | Intron | NP_001275572.1 | ||||
NM_001288644.1 | Intron | NP_001275573.1 | ||||
NM_001288645.1 | Intron | NP_001275574.1 | ||||
NM_001288646.1 | Intron | NP_001275575.1 | ||||
NM_001288647.1 | Intron | NP_001275576.1 | ||||
NM_001288648.1 | Intron | NP_001275577.1 | ||||
NM_001288649.1 | Intron | NP_001275578.1 | ||||
NM_020652.2 | Intron | NP_065703.1 |