Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCCGGCCCACAATGAAGATCTG[A/G]GAGGCAGGCAGGCCCAGCACGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608921 | ||||||||||||||||||||
Literature Links: |
FAM64A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM64A - family with sequence similarity 64 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITPNM3 - PITPNM family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165966.1 | 2719 | Silent Mutation | TCC,TCT | S,S 820 | NP_001159438.1 | |
NM_031220.3 | 2719 | Silent Mutation | TCC,TCT | S,S 856 | NP_112497.2 | |
XM_011524015.2 | 2719 | Intron | XP_011522317.1 | |||
XM_011524016.2 | 2719 | Intron | XP_011522318.1 | |||
XM_011524017.2 | 2719 | Intron | XP_011522319.1 |