Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCGCTTCTTGCCACTGCTGACG[C/T]GGATGAAGCCGTCAGCATCCTTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180466 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPL19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPL19 - ribosomal protein L19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAC2 - SH3 and cysteine rich domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198993.3 | 1313 | Missense Mutation | NP_945344.1 | |||
XM_017024580.1 | 1313 | Missense Mutation | XP_016880069.1 | |||
XM_017024581.1 | 1313 | Missense Mutation | XP_016880070.1 | |||
XM_017024582.1 | 1313 | Intron | XP_016880071.1 | |||
XM_017024583.1 | 1313 | Intron | XP_016880072.1 |