Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCACTGGGGAGAGAGCTTCTCCT[C/T]CAGGACGCCGTCGGGGGCGCACACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607211 MIM: 605270 MIM: 610117 | ||||||||||||||||||||
Literature Links: |
CARD14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CARD14 - caspase recruitment domain family member 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371920 - uncharacterized LOC105371920 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SGSH - N-sulfoglucosamine sulfohydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000199.3 | 2394 | Missense Mutation | AAG,GAG | K,E 488 | NP_000190.1 | |
XM_005257582.3 | 2394 | UTR 3 | XP_005257639.1 | |||
XM_005257583.4 | 2394 | Intron | XP_005257640.1 | |||
XM_011525126.1 | 2394 | Intron | XP_011523428.1 | |||
XM_011525127.2 | 2394 | UTR 3 | XP_011523429.1 | |||
XM_017024951.1 | 2394 | Intron | XP_016880440.1 | |||
XM_017024952.1 | 2394 | UTR 3 | XP_016880441.1 |
SLC26A11 - solute carrier family 26 member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |