Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGTTTGGGTACAACATTGGGGT[C/T]ATCAATGCCCCTCAGAAGGTGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138190 MIM: 611447 | ||||||||||||||||||||
Literature Links: |
SLC2A4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC2A4 - solute carrier family 2 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042.2 | 332 | Silent Mutation | GTC,GTT | V,V 44 | NP_001033.1 |
YBX2 - Y-box binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |