Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCTCTGGCCTCCTCAGCTGTAA[C/T]CTCCTGGGAGTAGAGGTCAGTGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610904 MIM: 611362 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNF8 - SNF8, ESCRT-II complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317192.1 | 847 | Missense Mutation | ATT,GTT | I,V 247 | NP_001304121.1 | |
NM_001317193.1 | 847 | Missense Mutation | ATT,GTT | I,V 231 | NP_001304122.1 | |
NM_001317194.1 | 847 | Missense Mutation | ATT,GTT | I,V 144 | NP_001304123.1 | |
NM_007241.3 | 847 | Missense Mutation | ATT,GTT | I,V 248 | NP_009172.2 | |
XM_017024075.1 | 847 | Missense Mutation | ATT,GTT | I,V 144 | XP_016879564.1 |
UBE2Z - ubiquitin conjugating enzyme E2 Z | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |