Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGATAGAGGGTAAACGTCTGCCA[C/T]GGGCAGCTTATAGGTTAACAAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612504 MIM: 612507 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA6 - ATP binding cassette subfamily A member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080284.2 | 5135 | Missense Mutation | ATG,GTG | M,V 1550 | NP_525023.2 | |
XM_011524574.2 | 5135 | Missense Mutation | ATG,GTG | M,V 1233 | XP_011522876.1 | |
XM_017024404.1 | 5135 | Missense Mutation | ATG,GTG | M,V 1550 | XP_016879893.1 | |
XM_017024405.1 | 5135 | Missense Mutation | ATG,GTG | M,V 1550 | XP_016879894.1 | |
XM_017024406.1 | 5135 | Missense Mutation | ATG,GTG | M,V 1355 | XP_016879895.1 | |
XM_017024407.1 | 5135 | Missense Mutation | ATG,GTG | M,V 1355 | XP_016879896.1 | |
XM_017024408.1 | 5135 | Missense Mutation | ATG,GTG | M,V 865 | XP_016879897.1 |
ABCA9 - ATP binding cassette subfamily A member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |