Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGGTCCAGCAGCCCTTTCCTGA[G/T]CAAGCGGTCTCTGAGTCGGAGTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615514 | ||||||||||||||||||||
Literature Links: |
CDK12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK12 - cyclin dependent kinase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015083.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | NP_055898.1 | |
NM_016507.3 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | NP_057591.2 | |
XM_005257456.3 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_005257513.1 | |
XM_005257458.4 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_005257515.1 | |
XM_011524892.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523194.1 | |
XM_011524893.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523195.1 | |
XM_011524894.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523196.1 | |
XM_011524895.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523197.1 | |
XM_011524896.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523198.1 | |
XM_011524897.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523199.1 | |
XM_011524898.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523200.1 | |
XM_011524899.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523201.1 | |
XM_011524900.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523202.1 | |
XM_011524901.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523203.1 | |
XM_011524902.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523204.1 | |
XM_011524903.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523205.1 | |
XM_011524905.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523207.1 | |
XM_011524906.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523208.1 | |
XM_011524907.2 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_011523209.1 | |
XM_017024744.1 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_016880233.1 | |
XM_017024745.1 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_016880234.1 | |
XM_017024746.1 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_016880235.1 | |
XM_017024747.1 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_016880236.1 | |
XM_017024748.1 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_016880237.1 | |
XM_017024749.1 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_016880238.1 | |
XM_017024750.1 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_016880239.1 | |
XM_017024751.1 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_016880240.1 | |
XM_017024752.1 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_016880241.1 | |
XM_017024753.1 | 1574 | Missense Mutation | AGC,ATC | S,I 338 | XP_016880242.1 |