Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAACTGCCATCCTGAAAACAAACA[C/T]GGTGAAAGTGCCCAAGCCCATCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611683 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FN3KRP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FN3KRP - fructosamine 3 kinase related protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024619.3 | 250 | Missense Mutation | ACG,ATG | T,M 67 | NP_078895.2 |
LOC101929552 - uncharacterized LOC101929552 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |