Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCATTGCCACCCTGGCCGTGAAG[A/C]GGGTAAGGCCAAGTGGGCGGGTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600362 MIM: 615498 MIM: 601243 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLII PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLII - FLII, actin remodeling protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIEF2 - mitochondrial elongation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144900.1 | 356 | Silent Mutation | AGG,CGG | R,R 49 | NP_001138372.1 | |
NM_139162.3 | 356 | Silent Mutation | AGG,CGG | R,R 49 | NP_631901.2 | |
NM_148886.1 | 356 | Silent Mutation | AGG,CGG | R,R 60 | NP_683684.2 | |
XM_017024190.1 | 356 | Silent Mutation | AGG,CGG | R,R 56 | XP_016879679.1 |
TOP3A - topoisomerase (DNA) III alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |