Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGACAGGTGCCATGTGTCTGTGC[C/T]GTGCGACATATGAGGAGCAGCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604375 MIM: 602375 MIM: 606794 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HGS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HGS - hepatocyte growth factor-regulated tyrosine kinase substrate | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL12 - mitochondrial ribosomal protein L12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002949.3 | 231 | Silent Mutation | GCC,GCT | A,A 33 | NP_002940.2 |
SLC25A10 - solute carrier family 25 member 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |