Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATGCTGCTGCCCCAGCCTTCCC[A/G]CAGACTTGGCCTAATGAAAATACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605398 MIM: 612383 MIM: 614312 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXCL16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXCL16 - C-X-C motif chemokine ligand 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100812.1 | 747 | Intron | NP_001094282.1 | |||
NM_022059.3 | 747 | Intron | NP_071342.2 |
MED11 - mediator complex subunit 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001683.3 | 747 | UTR 3 | NP_001001683.1 | |||
NM_001305000.1 | 747 | UTR 3 | NP_001291929.1 |
ZMYND15 - zinc finger MYND-type containing 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |